missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromonaphthalene, 98%

Product Code. 11473687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11473687 5 g 5g
11483687 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11473687

Brand: Thermo Scientific Alfa Aesar A18171.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83-53-4 | C10H6Br2 | 285.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-53-4
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD00041823
InChI Key IBGUDZMIAZLJNY-UHFFFAOYSA-N
Synonym naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g
PubChem CID 66521
IUPAC Name 1,4-dibromonaphthalene
SMILES BrC1=C2C=CC=CC2=C(Br)C=C1

Specifications

Melting Point 80°C to 82°C
Quantity 5 g
Formula Weight 285.97
Percent Purity 98%
Chemical Name or Material 1,4-Dibromonaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.