missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobutane, 97%

Product Code. p-7025700
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11479257

Brand: Thermo Scientific Alfa Aesar B21337.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-56-5 | C4H8Cl2 | 127.01 g/mol

Used in organic synthesis including adiponitrile. It is also used as an active pharmaceutical intermediate. 1,4-dichlorobutane (1,4-DCB) can be used as an organic solvent (O) for ion-transfer voltammetry at water (W) interface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in organic synthesis including adiponitrile. It is also used as an active pharmaceutical intermediate. 1,4-dichlorobutane (1,4-DCB) can be used as an organic solvent (O) for ion-transfer voltammetry at water (W) interface.

Solubility
Soluble in water at 20°C2.4g/L.

Notes
Store away from oxidizing agents, alkali metals and alkali earth metals. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-56-5
Molecular Formula C4H8Cl2
Molecular Weight (g/mol) 127.01
MDL Number MFCD00001011
InChI Key KJDRSWPQXHESDQ-UHFFFAOYSA-N
Synonym butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc
PubChem CID 8059
IUPAC Name 1,4-dichlorobutane
SMILES ClCCCCCl

Specifications

Melting Point -38°C
Density 1.139
Boiling Point 155°C
Flash Point 40°C (104°F)
Odor Aromatic
Refractive Index 1.454
Quantity 250 g
UN Number UN1993
Beilstein 635673
Solubility Information Soluble in water at 20°C2.4g/L.
Formula Weight 127.01
Percent Purity 97%
Chemical Name or Material 1,4-Dichlorobutane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.