missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals

Product Code. 10646682 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
250mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646682 1 L Glass bottle 1 L
10530551 5 mL Glass bottle 5mL
10000860 100 mL Glass bottle 100mL
10315750 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10646682 Supplier Thermo Scientific Acros Supplier No. 113830010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-69-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00001975
InChI Key BTGRAWJCKBQKAO-UHFFFAOYSA-N
Synonym adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
PubChem CID 8128
IUPAC Name hexanedinitrile
SMILES C(CCC#N)CC#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 1.0°C to 3.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 295.0°C
Flash Point 163°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NC(CH2)4CN
Refractive Index 1.4370 to 1.4390
Quantity 1 L
Beilstein 02, 653
Merck Index 15, 152
Specific Gravity 0.95
Solubility Information Solubility in water: 90g/L (20°C). Other solubilities: soluble in alcohol and chloroform
Viscosity 5.8 mPa.s (30°C)
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 4-Dicyanobutane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Dicyanobutane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.