missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodobutane, 99%, stab. with copper

Product Code. 11413317
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413317 100 g 100g
11403317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413317 Supplier Thermo Scientific Alfa Aesar Supplier No. A16216.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 628-21-7 | C4H8I2 | 309.92 g/mol

1,4-Diiodobutane is used as an additive to enhance perovskite solar cell performance. It is used as a crosslinking agent for the synthesis of anion exchange membranes from cellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diiodobutane is used as an additive to enhance perovskite solar cell performance. It is used as a crosslinking agent for the synthesis of anion exchange membranes from cellulose.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-21-7
Molecular Formula C4H8I2
Molecular Weight (g/mol) 309.92
MDL Number MFCD00001099
InChI Key ROUYUBHVBIKMQO-UHFFFAOYSA-N
Synonym tetramethylene iodide, butane, 1,4-diiodo, tetramethylene diiodide, 1,4-diodobutane, a,?-diiodobutane, 1,4 diiodobutane, butane,4-diiodo, 1,4-diiodo-butane, pubchem3842, 1,4-bis iodanyl butane
PubChem CID 12337
IUPAC Name 1,4-diiodobutane
SMILES ICCCCI

Specifications

Melting Point 5°C to 7°C
Density 2.35
Boiling Point 108°C to 110°C (10 mmHg)
Refractive Index 1.6212
Quantity 100 g
Beilstein 1098276
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 309.92
Percent Purity 99%
Chemical Name or Material 1,4-Diiodobutane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.