missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodooctafluorobutane, 97%, stab. with copper

Product Code. 11495277
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495277 25 g 25g
11485277 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495277 Supplier Thermo Scientific Alfa Aesar Supplier No. B22660.14
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 375-50-8 | C4F8I2 | 453.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-50-8
Molecular Formula C4F8I2
Molecular Weight (g/mol) 453.84
MDL Number MFCD00042263
InChI Key JILAKKYYZPDQBE-UHFFFAOYSA-N
Synonym 1,4-diiodooctafluorobutane, octafluoro-1,4-diiodobutane, 1,4-diiodoperfluorobutane, perfluoro-1,4-diiodobutane, 1,4-diiodo octafluorobutane, butane, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo, acmc-1aigi, i cf2 4i, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo-butane
PubChem CID 78993
IUPAC Name 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodobutane
SMILES FC(F)(I)C(F)(F)C(F)(F)C(F)(F)I

Specifications

Melting Point -9°C
Density 2.487
Boiling Point 85°C (100 mmHg)
Linear Formula I(CF2)4I
Refractive Index 1.4294
Quantity 25 g
Beilstein 1777548
Sensitivity Light sensitive
Formula Weight 453.84
Percent Purity 97%
Chemical Name or Material 1,4-Diiodooctafluorobutane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.