missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diisopropylbenzene, 98%

Product Code. 11369789
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369789 500 g 500g
11349789 25 g 25g
11359789 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369789 Supplier Thermo Scientific Alfa Aesar Supplier No. B22871.36
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 100-18-5 | C12H18 | 162.28 g/mol

1,4-Diisopropylbenzene is used as an intermediate for the synthesis of stabilizers, polymers, synthetic lubricants and hydroperoxides. It is also used in the trivalent actinide-lanthanide separation by phosphorus reagent extraction from aqueous komplexes (TALSPEAK) process for separating trivalent lanthanides from trivalent actinide cations. It finds application in nuclear fuel reprocessing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diisopropylbenzene is used as an intermediate for the synthesis of stabilizers, polymers, synthetic lubricants and hydroperoxides. It is also used in the trivalent actinide-lanthanide separation by phosphorus reagent extraction from aqueous komplexes (TALSPEAK) process for separating trivalent lanthanides from trivalent actinide cations. It finds application in nuclear fuel reprocessing.

Solubility
Soluble in alcohol, ether, acetone and benzene. Insoluble in water.

Notes
Incompatible with oxidizing agents and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-18-5
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00008892
InChI Key SPPWGCYEYAMHDT-UHFFFAOYSA-N
Synonym 1,4-diisopropylbenzene, p-diisopropylbenzene, benzene, p-diisopropyl, benzene, 1,4-bis 1-methylethyl, para-diisopropylbenzene, p-diisopropylbenzol, 1,4-bis 1-methylethyl benzene, unii-5m62031nzp, 1,4-di propan-2-yl benzene, dsstox_cid_6652
PubChem CID 7486
IUPAC Name 1,4-di(propan-2-yl)benzene
SMILES CC(C)C1=CC=C(C=C1)C(C)C

Specifications

Melting Point -17°C
Density 0.857
Boiling Point 210°C
Flash Point 76°C (170°F)
Odor Aromatic
Refractive Index 1.489
Quantity 500 g
Beilstein 1854739
Solubility Information Soluble in alcohol,ether,acetone and benzene. Insoluble in water.
Formula Weight 162.27
Percent Purity 99%
Chemical Name or Material 1,4-Diisopropylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.