missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxybenzene, 98%, BLD Pharm™

Product Code. 30784403
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30784403 100 g 100g
30777726 1 kg 1kg
30778095 25 g 25g
30774497 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30784403 Supplier BLD Pharm Supplier No. BD134163100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 150-78-7
Molecular Formula C8H10O2
MDL Number MFCD00008401
Quantity 100 g
SMILES COC1=CC=C(OC)C=C1
Molecular Weight (g/mol) 138.16
Percent Purity 98%
Chemical Name or Material 1,4-Dimethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.