missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethyl-2-iodobenzene 98%, ACROS Organics™

Product Code. 10668814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10668814 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10668814 Supplier Thermo Scientific Acros Supplier No. 393250050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1122-42-5
CAS Min % 97.5
Color Yellow
CAS Max % 100.0
Density 1.6170g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5960 to 1.5980
Assay Percent Range 98%
Specific Gravity 1.617
Molecular Formula C8H9I
Linear Formula IC6H3(CH3)2
MDL Number MFCD00013708
Synonym 2-iodo-p-xylene, 1,4-dimethyl-2-iodobenzene, p-xylene, 2-iodo, 1-iodo-2,5-dimethylbenzene, benzene, 2-iodo-1,4-dimethyl, 2-iodo-4-xylene, 2,5-dimethyliodobenzene, 2-iodo-1,4-dimethyl-benzene, 2,5-dimethyl-1-iodobenzene, pubchem3875
Boiling Point 229.0°C to 260.0°C
InChI Key WYZVNUSNUCABRF-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)C)I
IUPAC Name 2-iodo-1,4-dimethylbenzene
Molecular Weight (g/mol) 232.064
PubChem CID 70731
Formula Weight 232.06
Percent Purity 98%
Chemical Name or Material 1, 4-Dimethyl-2-iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Dimethyl-2-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.