missing translation for 'onlineSavingsMsg'
Learn More

1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, Thermo Scientific™

Product Code. 10892611
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10892611 250 mg Amber Glass Bottle 250mg
10724482 1 g Amber Glass Bottle 1g
10526820 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10892611 Supplier Thermo Scientific Maybridge Supplier No. CC13851CB
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1174064-55-1
Molecular Formula C7H16Cl2N2O2
Molecular Weight (g/mol) 231.117
InChI Key UPJQXMOMHHDOSZ-UHFFFAOYSA-N
Synonym 1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid,dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid, chloride, chloride, 1,4-dimethylpiperazine-2-carboxylic acid-hydrogen chloride 1/2
PubChem CID 45594293
IUPAC Name 1,4-dimethylpiperazine-2-carboxylic acid;dihydrochloride
SMILES CN1CCN(C(C1)C(=O)O)C.Cl.Cl

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 231.12
Percent Purity 95%
Chemical Name or Material 1, 4-dimethylpiperazine-2-carboxylic acid dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.