missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane-2,5-dione, 97%, Thermo Scientific™

Product Code. 10223612
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10223612 10 g Amber Glass Bottle 10g
10101132 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10223612 Supplier Thermo Scientific Maybridge Supplier No. SB01616EA
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 02-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 502-97-6
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00081108
InChI Key RKDVKSZUMVYZHH-UHFFFAOYSA-N
Synonym glycolide, glycollide, unii-yrz676pgu6, p-dioxane-2,5-dione, yrz676pgu6, acmc-1aybf, 1,4-dioxan-2,5-dione, 2,5-dioxo-1,4-dioxane, p-dioxane-2,5-dione 8ci
PubChem CID 65432
IUPAC Name 1,4-dioxane-2,5-dione
SMILES O=C1COC(=O)CO1

Specifications

CAS Min % 97.0
Quantity 10 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Formula Weight 116.07
Percent Purity 97%
Chemical Name or Material 1, 4-Dioxane-2, 5-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.