missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, 99.5%, for HPLC
GREENER_CHOICE

Product Code. 10613722 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10613722 1 L Glass Bottle 1 L
10267210 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10613722 Supplier Thermo Scientific Supplier No. 268340010
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-91-1, C4H8O2, 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 12.0°C
Absorbance 1 cm cell vs HPLC-grade water, 1 cm cell vs HPLC-grade water at 0 nm, 0.03 max. at 270nm, 0.05 max. at 260nm, 0.075 max. at 250nm, 0.125 max. at 240nm, 0.22 max. at 230nm, 0.4 max. at 220nm, 1 max. at 218nm
CAS Min % 99.4
Color Brown to Yellow
CAS Max % 100.0
Density 1.0340g/mL
Quantity 1 L
Flash Point 12°C
Packaging Glass Bottle
Physical Form Solution
Refractive Index 1.4205 to 1.4220
Residue after Evaporation 0.0002 % max.
Assay Percent Range 99.5%
Specific Gravity 1.034
Viscosity 1.32 mPa.s (20°C)
Water 0.025% Max.
Beilstein 19, 3
Fieser 01,333
Merck Index 15, 3328
Green Features ACT Ecolabel (Silver)
Boiling Point 101.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
Formula Weight 88.11
Percent Purity 99.5%
Grade HPLC
Chemical Name or Material 1, 4-Dioxane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Dioxane
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H350-May cause cancer.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-Dioxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.