missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™

Product Code. 10783203 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10783203 1 L 1 L
10061931 100 mL 100mL
10368143 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10783203 Supplier Thermo Scientific Acros Supplier No. 326890010
  • ONLINE EXCLUSIVE PRICE
    150.00 €
    145.00 € / 1 L
    Save 5.00 €
    3% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 12.0°C
CAS Min % 99.75
CAS Max % 100.0
Density 1.0340g/mL
Quantity 1 L
Flash Point 12°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4205 to 1.4220
Residue after Evaporation 0.001 % max.
Assay Percent Range 99.8%
Specific Gravity 1.034
Viscosity 1.32 mPa.s (20°C)
UN Number 1165
Water 0.005% Max. (Coulometric)
Beilstein 19, 3
Fieser 01,333
Merck Index 15, 3328
Boiling Point 101.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
Formula Weight 88.11
Percent Purity 99.8%
Grade Extra Dry
Chemical Name or Material 1,4-Dioxane, Water <50ppm
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Dioxane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.