missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diphenylbutadiyne, 99%

Product Code. 11475543
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11475543 100 g 100g
11455543 5 g 5g
11465543 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11475543

Brand: Thermo Scientific Alfa Aesar A13471.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 886-66-8 | C16H10 | 202.26 g/mol

1,4-Diphenylbutadiyne was used in the preparation of 7,8-dehydropurpurin dimers via two-fold Pd-catalyzed [3+2] annulation of meso-bromoporphyrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diphenylbutadiyne was used in the preparation of 7,8-dehydropurpurin dimers via two-fold Pd-catalyzed [3+2] annulation of meso-bromoporphyrin.

Solubility
Sparingly soluble in water(5.5E-4 g/L) (25°C). Soluble in toluene almost transparency.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-66-8
Molecular Formula C16H10
Molecular Weight (g/mol) 202.26
MDL Number MFCD00004787
InChI Key HMQFJYLWNWIYKQ-UHFFFAOYSA-N
Synonym 1,4-diphenylbutadiyne, diphenylbutadiyne, diphenyldiacetylene, butadiyne, diphenyl, diphenyl-1,3-butadiyne, benzene, 1,1'-1,3-butadiyne-1,4-diyl bis, 1,4-diphenyl-1,3-butadiyne, 1,4-diphenylbuta-1,3-diyne, 1,1'-buta-1,3-diyne-1,4-diyldibenzene, 4-phenyl-1,3-butadiynyl benzene
PubChem CID 70174
ChEBI CHEBI:51588
IUPAC Name 4-phenylbuta-1,3-diynylbenzene
SMILES C1=CC=C(C=C1)C#CC#CC1=CC=CC=C1

Specifications

Melting Point 86°C to 87°C
Quantity 100 g
Beilstein 1910105
Solubility Information Sparingly soluble in water(5.5E-4g/L) (25°C). Soluble in toluene almost transparency.
Formula Weight 202.26
Percent Purity 99%
Chemical Name or Material 1,4-Diphenylbutadiyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.