missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dithiothreitol, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15568774 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
POLYSTORMOR
Poly Bottle
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15568774 100 g Poly Bottle Each
15548774 5 g POLYSTORMOR Each
15558774 25 g POLYSTORMOR Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15568774 Supplier Avantor J.T.Baker Supplier No. F78003
  • ONLINE EXCLUSIVE PRICE
    2758.35 €
    1836.63 € / Pack of 1
    Save 921.72 €
    33% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

  • High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography.
  • For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 12-3-3483
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S

Specifications

Flash Point 109.00°C
Packaging Poly Bottle
Quantity 100 g
Specific Gravity 1L = 1.00kg
Formula Weight 154.25
Grade Ultrapure Bioreagent
Chemical Name or Material 1,4-Dithiothreitol
compliance-icons
Product Identifier
  • Dithiothreitol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Dithiothreitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.