missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10364562
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10364562 1 g 1g
10577264 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10364562 Supplier Thermo Scientific Maybridge Supplier No. CC00119DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 423768-49-4
Molecular Formula C11H8ClFN2O
Molecular Weight (g/mol) 238.646
MDL Number MFCD03659694
InChI Key INIAJXQDJISNJB-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methylpyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methyl-4-pyrazolecarbonyl chloride, 1-4-fluorophenyl-5-methyl-pyrazole-4-carbonyl chloride
PubChem CID 2779893
IUPAC Name 1-(4-fluorophenyl)-5-methylpyrazole-4-carbonyl chloride
SMILES CC1=C(C=NN1C2=CC=C(C=C2)F)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 238.65
Percent Purity 97%
Chemical Name or Material 1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.