Learn More
Trajan Scientific and Medical™ 1/4 in. Graphite Fittings
Product Code. 15208873
Change view
Click to view available options
Diameter (Metric) Inside:
1/8 in.
5/16 in.
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside unitSize
15208873 5/16 in. Pack of 10
15298863 1/8 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15208873 Supplier Trajan Scientific and Medical™ Supplier No. 072617
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Provide leak-free system connections. Trajan Scientific and Medical 1/4 in. Graphite Fittings leads to improved chromatography for non-MS systems.
  • Easy to use
  • Forms reliable seal
  • Reusable
  • Soft material, forms soft grip to the capillary column
  • Low emissions
Specifications
Type Graphite Reducing Ferrule 1/4 in. Fitting 5/16 in. x 9.5mm x 4.6mm
Size (Metric) Fitting 1/4 in.
Size (Metric) 9.5 mm x 4.6 mm
Material Graphite
Quantity 10/Pk.
Diameter (Metric) Inside 5/16 in.