missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Iodophenyl)pyrrole, 97%

Product Code. 11488176
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488176 1 g 1g
11498176 5 g 5g
11408186 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488176 Supplier Thermo Scientific Alfa Aesar Supplier No. A15305.03
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 92636-36-7 | C10H8IN | 269.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92636-36-7
Molecular Formula C10H8IN
Molecular Weight (g/mol) 269.085
MDL Number MFCD00052399
InChI Key FMURNAZHVQDQQN-UHFFFAOYSA-N
Synonym 1-4-iodophenyl pyrrole, 1-4-iodophenyl-1h-pyrrole, acmc-20apaj, 4-iodophenyl pyrrole, maybridge1_002926, 1-4-iodo-phenyl-1h-pyrrole, 1h-pyrrole,1-4-iodophenyl, 1-4-iodophenyl-1h-pyrrole #
PubChem CID 272430
IUPAC Name 1-(4-iodophenyl)pyrrole
SMILES C1=CN(C=C1)C2=CC=C(C=C2)I

Specifications

Melting Point 128°C to 133°C
Quantity 1 g
Beilstein 7915717
Sensitivity Light sensitive
Formula Weight 269.09
Percent Purity 97%
Chemical Name or Material 1-(4-Iodophenyl)pyrrole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.