missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-6,7-dihydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic Acid, TRC

Product Code. 30474342
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474342 25 mg 25mg
30466654 10 mg 10mg
30470717 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474342 Supplier Toronto Research Chemicals Supplier No. TRCM28708525MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2518444-61-4
Molecular Formula C25H22N4O5
Molecular Weight (g/mol) 458.47
IUPAC Name 1-(4-methoxyphenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-6,7-dihydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic acid
SMILES OC(C1=NN(C2=CC=C(OC)C=C2)C(C3=O)=C1C=CN3C(C=C4)=CC=C4N5C(CCCC5)=O)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H22N4O5/c1-34-19-11-9-18(10-12-19)29-23-20(22(26-29)25(32)33)13-15-28(24(23)31)17-7-5-16(6-8-17)27-14-3-2-4-21(27)30/h5-13,15H,2-4,14H2,1H3,(H,32,33)
Formula Weight 458.16
Chemical Name or Material 1-(4-Methoxyphenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-6,7-dihydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.