missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)piperazine, 96%, Thermo Scientific™

Product Code. 10718312 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10718312 250 g Glass bottle 250g
10033240 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10718312 Supplier Thermo Scientific Acros Supplier No. 255832500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 38212-30-5
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.26
MDL Number MFCD00040736
InChI Key MRDGZSKYFPGAKP-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl piperazine, n-4-methoxyphenyl piperazine, n-p-methoxyphenyl piperazine, piperazine, 1-4-methoxyphenyl, unii-p385m92gyg, 4-methoxyphenylpiperazine, 1-4-anisyl piperazine, chembl59517, 1-4-methoxyphenyl-piperazine, 1-4-methoxy-phenyl-piperazine
PubChem CID 269722
IUPAC Name 1-(4-methoxyphenyl)piperazine
SMILES COC1=CC=C(C=C1)N1CCNCC1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Amber to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 250 g
Formula Weight 192.25
Percent Purity 96%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 1-(4-Methoxyphenyl)piperazine, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(4-Methoxyphenyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.