missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methylphenyl)-1-cyclopropanecarboxylic acid, 98%

Product Code. 10265840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10265840 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10265840 Supplier Thermo Scientific Acros Supplier No. 170700100
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 83846-66-6 | C11H12O2 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 83846-66-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 109°C to 114°C
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C11H12O2
MDL Number MFCD00066918
Quantity 10 g
Synonym 1-p-tolyl cyclopropanecarboxylic acid, 1-4-methylphenyl cyclopropanecarboxylic acid, 1-4-methylphenyl-1-cyclopropanecarboxylic acid, 1-4-methylphenyl cyclopropane-1-carboxylic acid, 1-p-tolyl-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-4-methylphenyl, 1-p-tolylcyclopropanecarboxylic acid, 1-4-methylphenyl-1-cyclopropan carboxylic acid, cyclopropanecarboxylicacid, 1-4-methylphenyl, 1-4-methylphenyl-1-cyclopropane carboxylic acid
InChI Key AYUGAOYMYXSOKU-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)C2(CC2)C(=O)O
IUPAC Name 1-(4-methylphenyl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol) 176.21
PubChem CID 98639
Formula Weight 176.21
Percent Purity 98%
Chemical Name or Material 1-(4-Methylphenyl)-1-cyclopropanecarboxylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-(4-Methylphenyl)-1-cyclopropanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.