missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Morpholinyl)-2-propanol, 98%

Product Code. 11392667
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392667 10 g 10g
11302677 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392667 Supplier Thermo Scientific Alfa Aesar Supplier No. L11859.09
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2109-66-2 | C7H15NO2 | 145.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2109-66-2
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD00023378
InChI Key YAXQOLYGKLGQKA-UHFFFAOYSA-N
Synonym n-2-hydroxypropyl morpholine, 1-morpholin-4-yl propan-2-ol, 1-morpholinopropan-2-ol, 1-morpholino-2-propanol, 4-2-hydroxypropyl morpholine, 1-4-morpholino-2-propanol, 4-morpholineethanol, alpha-methyl, 4-morpholineethanol, .alpha.-methyl, acmc-1ck5f, 1-morpholino propan-2-ol
PubChem CID 97957
IUPAC Name 1-morpholin-4-ylpropan-2-ol
SMILES CC(CN1CCOCC1)O

Specifications

Density 1.02
Boiling Point 218°C
Flash Point 92°C (197°F)
Refractive Index 1.4635
Quantity 10 g
Beilstein 1443
Formula Weight 145.2
Percent Purity 98%
Chemical Name or Material 1-(4-Morpholinyl)-2-propanol
compliance-icons
Product Identifier
  • 1-(4-Morpholinyl)-2-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-(4-Morpholinyl)-2-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.