missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)piperazine, 98%

Product Code. 11349475
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349475 100 g 100g
11329475 5 g 5g
11339475 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349475 Supplier Thermo Scientific Alfa Aesar Supplier No. L18700.22
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 6269-89-2 | C10H13N3O2 | 207.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6269-89-2
Molecular Formula C10H13N3O2
Molecular Weight (g/mol) 207.233
MDL Number MFCD00005961
InChI Key VWOJSRICSKDKAW-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl piperazine, 1-4-nitro-phenyl-piperazine, piperazine, 1-4-nitrophenyl, 1-4-nitrophenyl-piperazine, n-4-nitrophenyl piperazine, n-4-nitrophenyl-piperazine, 4-nitrophenyl piperazine, 1-4-nitrophenyl piperazin, pubchem8580, 4-nitrophenylpiperazine
PubChem CID 80447
SMILES C1CN(CCN1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 129°C to 133°C
Quantity 100 g
UN Number UN2811
Formula Weight 207.23
Percent Purity ≥98%
Chemical Name or Material 1-(4-Nitrophenyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.