missing translation for 'onlineSavingsMsg'
Learn More

1.4-Phenylenediacetic acid, 97%

Product Code. 10362083 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10362083 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10362083 Supplier Thermo Scientific Acros Supplier No. 130510050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7325-46-4 | C10H8O4 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7325-46-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 250.0°C to 256.0°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C10H8O4
Linear Formula C6H4(CH2CO2H)2
MDL Number MFCD00004354
Quantity 5 g
Beilstein 09, 875
Synonym 1,4-phenylenediacetic acid, p-phenylenediacetic acid, 2,2'-1,4-phenylene diacetic acid, 1,4-benzenediacetic acid, 2-4-carboxymethyl phenyl acetic acid, 4-carboxymethyl phenyl acetic acid, p-benzenediacetic acid, acmc-209oq4, 1.4-phenylenediacetic acid, ksc378o7r
InChI Key SLWIPPZWFZGHEU-UHFFFAOYSA-L
SMILES [O-]C(=O)CC1=CC=C(CC([O-])=O)C=C1
IUPAC Name 2-[4-(carboxylatomethyl)phenyl]acetate
Molecular Weight (g/mol) 192.17
PubChem CID 81760
Formula Weight 194.19
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1.4-Phenylenediacetic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1.4-Phenylenediacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.