missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylenediamine, TRC

Product Code. 30040800
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040800 1 g 1g
30009338 100 g 100g
30011782 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040800 Supplier Toronto Research Chemicals Supplier No. TRCP3198451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-50-3
Formula Weight 108.0687
InChI Formula InChI=1S/C6H8N2/c7-5-1-2-6(8)4-3-5/h1-4H,7-8H2
IUPAC Name benzene-1,4-diamine
Molecular Formula C6 H8 N2
Molecular Weight (g/mol) 108.14
Percent Purity >95%
SMILES Nc1ccc(N)cc1
Synonym 1,4-Benzenediamine, p-Phenylenediamine (8CI), (4-Aminophenyl)amine, 1,4-Diaminobenzene, 1,4-Diaminobenzol, 1,4-Phenylenediamine, 4-Aminoaniline, 4-Phenylenediamine, BASF Ursol D, Benzofur D, Black for Fur D, C.I. 76060, C.I. Developer 13, C.I. Oxidation Base 10, Developer PF, Durafur Black R, Fouramine D, Fourrine 1, Fourrine D, Fur Black 41867, Fur Brown 41866, Fur Yellow, Furro D, Futramine D, NSC 4777, Nako H, Orsin, PDA, PPD, Paramine, Pelagol D, Pelagol DR, Pelagol Grey D, Peltol D, Renal PF, Rodol D, Rodol DJ, Tertral D, Ursol D, Zoba Black D, p-Aminoaniline, p-Benzenediamine, p-Diaminobenzene, pPDA
Chemical Name or Material 1,4-Phenylenediamine (1,4-Diaminobenzene)
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.