missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Pyridyl)ethylamine, 97%

Product Code. 11419460
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419460 250 mg 250mg
11429460 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419460 Supplier Thermo Scientific Alfa Aesar Supplier No. H50303.MD
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50392-78-4 | C7H11N2 | 123.18 g/mol

1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50392-78-4
Molecular Formula C7H11N2
Molecular Weight (g/mol) 123.18
MDL Number MFCD02245004,MFCD09256818,MFCD09256819
InChI Key HIZMJYQEHFJWQY-LURJTMIESA-O
Synonym 1-4-pyridyl ethylamine, 1-pyridin-4-yl-ethylamine, 1-pyridin-4-yl ethanamine, 1-4-pyridinyl ethanamine, 1-pyridin-4-yl ethan-1-amine, 1-4-pyridinyl ethanamin, 4-1-aminoethyl pyridine, alpha-methyl-4-pyridinemethylamine, 1-pyridin-4-ylethylamine, acmc-1aq0e
PubChem CID 247998
IUPAC Name 1-pyridin-4-ylethanamine
SMILES C[C@H]([NH3+])C1=CC=NC=C1

Specifications

Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in water
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 1-(4-Pyridyl)ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.