missing translation for 'onlineSavingsMsg'
Learn More

1,5-Anhydro-D-sorbitol, 97%

Product Code. 15559503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15559503 250 mg 250mg
15549503 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15559503 Supplier Thermo Scientific Acros Supplier No. 459912500
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 154-58-5 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-58-5
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
InChI Key MPCAJMNYNOGXPB-SLPGGIOYSA-N
Synonym 1,5-anhydro-d-glucitol, 1,5-anhydroglucitol, 1,5-anhydrosorbitol, 1,5-anhydro-d-sorbitol, 1-deoxy-d-glucopyranose, aceritol, 1-deoxy-d-glucose, d-glucitol, 1,5-anhydro, unii-54bb3b7xmz, glucitol, 1,5-anhydro
PubChem CID 64960
ChEBI CHEBI:16070
IUPAC Name (2R,3S,4R,5S)-2-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(C(C(C(O1)CO)O)O)O

Specifications

Melting Point 141.0°C to 144.0°C
Infrared Spectrum Authentic
Quantity 250 mg
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 1,5-Anhydro-D-sorbitol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.