missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Bromo-2,3-dihydro-1H-indol-1-yl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10235472
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10235472 1 g 1g
10743633 10 g 10g
10314933 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10235472 Supplier Thermo Scientific Maybridge Supplier No. KM08258DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 22190-38-1
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD00056017
InChI Key WQKQAIXOTCPWFE-UHFFFAOYSA-N
Synonym 1-acetyl-5-bromoindoline, 1-5-bromoindolin-1-yl ethanone, 1-5-bromo-2,3-dihydro-1h-indol-1-yl ethan-1-one, 1-5-bromo-2,3-dihydroindol-1-yl ethanone, 1-5-bromo-2,3-dihydro-1h-indol-1-yl ethanone, 1h-indole, 1-acetyl-5-bromo-2,3-dihydro, 1-acetyl-5-bromo-2,3-dihydroindole, acmc-20ailu, 5-bromo-n-acetylindoline, maybridge1_005011
PubChem CID 721847
IUPAC Name 1-(5-bromo-2,3-dihydroindol-1-yl)ethanone
SMILES CC(=O)N1CCC2=C1C=CC(=C2)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 240.1
Percent Purity 97%
Chemical Name or Material 1-(5-Bromo-2, 3-dihydro-1H-indol-1-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.