missing translation for 'onlineSavingsMsg'
Learn More

1,5-Cyclooctadiene, 99%, stabilized

Product Code. 10793141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10793141 250 mL Glass bottle 250mL
10627251 1 L Glass bottle 1 L
10013163 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10793141 Supplier Thermo Scientific Acros Supplier No. 297122500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 111-78-4 | C8H12 | 108.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-78-4
Molecular Formula C8H12
Molecular Weight (g/mol) 108.18
MDL Number MFCD00001752
InChI Key VYXHVRARDIDEHS-QGTKBVGQSA-N
Synonym 1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
PubChem CID 10937607
SMILES C1C\C=C/CC\C=C/1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -69.5°C
Color Colorless
Density 0.8800g/mL
Boiling Point 151°C (101.3 kPa)
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4920 to 1.4940
Quantity 250 mL
Beilstein 05, 116
Merck Index 15, 2730
Specific Gravity 0.88
Solubility Information Solubility in water: 780mg/l (20°C). Other solubilities: soluble in benzene and carbon tetrachloride
IUPAC Name (5Z)-cycloocta-1,5-diene
Stabilizer 50 to 150ppm t-butylcatechol
Viscosity 1.3 mPa.s (20°C)
Formula Weight 108.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 5-Cyclooctadiene, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,5-Cyclooctadiene, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.