missing translation for 'onlineSavingsMsg'
Learn More

(1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) Hexafluorophosphate, 97%, Thermo Scientific Chemicals

Product Code. 10756753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10756753 250 mg Glass bottle 250mg
10056172 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10756753

Brand: Thermo Scientific Acros 377942500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(1, 5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate, 97%, C34H38IrP2.F6P, CAS Number-38465-86-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38465-86-0
Molecular Formula C34H38F6IrP3
Molecular Weight (g/mol) 845.81
MDL Number MFCD00064800
InChI Key LXKHQEQLBSJJCO-JXNOXZOESA-N
Synonym 1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate
PubChem CID 5702663
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate
SMILES [Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 224.0°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass bottle
Quantity 250 mg
Formula Weight 845.81
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (1, 5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • (1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.