missing translation for 'onlineSavingsMsg'
Learn More

1,5-Cyclooctadiene(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate, 99%

Product Code. 11369717
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369717 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11369717 Supplier Thermo Scientific Alfa Aesar Supplier No. 042057.MF
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64536-78-3 | C31H50F6IrNP2 | 804.90 g/mol

1,5-Cyclooctadiene(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate is used as a catalyst for hydrogenation of mono-, di-, tri-, and tetra-substituted substrates and the isomerization and hydroboration of alkenes. It is also used in isotope exchange reactions, especially the direct exchange of a hydrogen atom with its isotopes deuterium and tritium. Carbonyl groups are also known to direct the hydrogenation by the Crabtree catalyst is highly regioselective.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Specifications

CAS 64536-78-3
Melting Point 175°C (decomposition)
Molecular Formula C31H50F6IrNP2
MDL Number MFCD00075097
Quantity 50 mg
Sensitivity Moisture sensitive
Synonym crabtree's catalyst, unii-816 rs2nbpn, 816 rs2nbpn, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium hexafluorophosphate, iridium i hexafluorophosphate, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium i hexafluorophosphate, tricyclohexylphosphine 1,5-cyclooctadiene pyridine iridium i hexafluorophosphate, felkin-crabtree catalyst, crabtree's catalyst mi, crabtree's catalyst hexafluorophosphate
Solubility Information Slightly soluble in acetone,dichloromethane,ethanol and diethyl ether; Insoluble aq. soln..
InChI Key UJXHUUQZACSUOG-KJWGIZLLSA-N
SMILES [Ir+].C1=CC=NC=C1.F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
IUPAC Name iridium(1+) (1Z,5Z)-cycloocta-1,5-diene hexafluoro-λ⁵-phosphanuide pyridine tricyclohexylphosphane
Molecular Weight (g/mol) 804.90
PubChem CID 5702647
Formula Weight 804.89
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Cyclooctadiene(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.