missing translation for 'onlineSavingsMsg'
Learn More

(1,5-Cyclooctadiene)rhodium(I) 2,4-pentanedionate

Product Code. 15457545
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457545 250 mg 250mg
11322039 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457545 Supplier Thermo Scientific Alfa Aesar Supplier No. H26083.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12245-39-5 | C13H19O2Rh | 310.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12245-39-5
Molecular Formula C13H19O2Rh
Molecular Weight (g/mol) 310.20
MDL Number MFCD00075046
InChI Key TVGPAHONTUMCNP-DWVXZKBMSA-M
Synonym acetylacetonato 1,5-cyclooctadiene rhodium i, rh acac cod, acetylacetonate 1,5-cyclooctadiene rhodium, acetylacetonato 1,5-cyclooctadiene rhodium, rhodium 1,5-cyclooctadiene-2,4-pentanedionate, acetylacetonato eta-cycloocta-1,5-diene rhodium i, 3z-4-rhodiooxy pent-3-en-2-one; 1,5-cyclooctadiene, z,z
PubChem CID 10935795
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;(Z)-4-hydroxypent-3-en-2-one;rhodium
SMILES [Rh+].C\C([O-])=C\C(C)=O.C1C\C=C/CC\C=C/1

Specifications

Melting Point 136°C to 140°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 310.19
Chemical Name or Material (1,5-Cyclooctadiene)rhodium(I) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.