missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminopentane, 98%

Product Code. 11456226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456226 5 g 5g
11466226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456226 Supplier Thermo Scientific Alfa Aesar Supplier No. B23039.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 462-94-2 | C5H14N2 | 102.18 g/mol

1,5-Diaminopentane is mainly used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diaminopentane is mainly used in organic synthesis.

Solubility
Soluble in water.

Notes
Air sensitive & hygroscopic. Incompatible with acid chlorides, acids, acid anhydrides, strong oxidizing agents, carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-94-2
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008239
InChI Key VHRGRCVQAFMJIZ-UHFFFAOYSA-N
Synonym 1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p
PubChem CID 273
ChEBI CHEBI:18127
IUPAC Name pentane-1,5-diamine
SMILES NCCCCCN

Specifications

Melting Point 9°C
Density 0.873
Boiling Point 178°C to 180°C
Assay Percent Range 0.98
Linear Formula H2N(CH2)5NH2
Refractive Index 1.4585
Quantity 5 g
UN Number UN2735
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,1609
Solubility Information Soluble in water.
Formula Weight 102.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 5-Diaminopentane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,5-Diaminopentane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Pentamethylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.