missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane, 97%

Product Code. 11409367
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409367 2 g 2g
11419367 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409367 Supplier Thermo Scientific Alfa Aesar Supplier No. B21405.04
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 371-83-5 | C5H4Br2F6 | 337.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-83-5
Molecular Formula C5H4Br2F6
Molecular Weight (g/mol) 337.89
MDL Number MFCD00236657
InChI Key XYMBGGTVDWPIBA-UHFFFAOYSA-N
Synonym pentane,1,5-dibromo-1,1,3,3,5,5-hexafluoro, 1,5-dibromo-1,1,3,3,5,5-hexafluoropentane
PubChem CID 2736777
IUPAC Name 1,5-dibromo-1,1,3,3,5,5-hexafluoropentane
SMILES FC(F)(Br)CC(F)(F)CC(F)(F)Br

Specifications

Density 2.003
Linear Formula BrCF2CH2CF2CH2CF2Br
Refractive Index 1.4632
Quantity 2 g
Formula Weight 337.89
Percent Purity 97%
Chemical Name or Material 1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.