missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromo-3-methylpentane, 98+%

Product Code. 11418887
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418887 5 g 5g
11428887 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418887 Supplier Thermo Scientific Alfa Aesar Supplier No. B20521.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 4457-72-1 | C6H12Br2 | 243.97 g/mol

1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 4457-72-1
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00039200
InChI Key YDPZWUMQKMLLHC-UHFFFAOYSA-N
Synonym 3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g
PubChem CID 138258
IUPAC Name 1,5-dibromo-3-methylpentane
SMILES CC(CCBr)CCBr

Specifica

Density 1.58
Boiling Point 233°C
Flash Point 101°C (214°F)
Linear Formula BrCH2CH2CH(CH3)CH2CH2Br
Refractive Index 1.508
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 243.98
Percent Purity ≥98%
Chemical Name or Material 1,5-Dibromo-3-methylpentane

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.