missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromopentane, 98%

Product Code. 11438387
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438387 100 g 100g
11448387 500 g 500g
11458387 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438387 Supplier Thermo Scientific Alfa Aesar Supplier No. A18465.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-24-0 | C5H10Br2 | 229.943 g/mol

1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.

Solubility
Miscible with water, slightly miscible with chloroform and benzene.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-24-0
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.943
MDL Number MFCD00000268
InChI Key IBODDUNKEPPBKW-UHFFFAOYSA-N
PubChem CID 8100
IUPAC Name 1,5-dibromopentane
SMILES C(CCBr)CCBr

Specifications

Melting Point -34°C
Density 1.688
Boiling Point 224°C
Flash Point 79°C (174°F)
Refractive Index 1.5122
Quantity 100 g
Beilstein 1209245
Synonym pentamethylene bromide, pentane, 1,5-dibromo, pentamethylene dibromide, unii-b8q228qys1, pentane-1,5-dibromide, 1,5-dibromo-pentane, 1,5-pentane dibromide, pentane,1,5-dibromo, br ch2 5br, ccris 8918
Solubility Information Miscible with water,slightly miscible with chloroform and benzene.
Formula Weight 229.95
Percent Purity 98%
Chemical Name or Material 1,5-Dibromopentane
Show More Show Less
compliance-icons
Product Identifier
  • 1,5-Dibromopentane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,5-Dibromopentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.