missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dichloropentane, 98%

Product Code. 11459643
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459643 25 g 25g
11469643 100 g 100g
11479643 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459643 Supplier Thermo Scientific Alfa Aesar Supplier No. A12433.14
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 628-76-2 | C5H10Cl2 | 141.035 g/mol

1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.

Solubility
Miscible with ethanol, ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-76-2
Molecular Formula C5H10Cl2
Molecular Weight (g/mol) 141.035
MDL Number MFCD00001017
InChI Key LBKDGROORAKTLC-UHFFFAOYSA-N
Synonym pentamethylene chloride, pentane, 1,5-dichloro, pentamethylene dichloride, amylene chloride, unii-6n4a5v83we, 1,5-dichlorobutane, pentane,5-dichloro, acmc-1ay6s, 1,5-bis chloranyl pentane, wln: g5g
PubChem CID 12353
IUPAC Name 1,5-dichloropentane
SMILES C(CCCl)CCCl

Specifications

Melting Point -72°C
Boiling Point 182°C
Flash Point 26°C (78°F)
Linear Formula Cl(CH2)5Cl
Quantity 25 g
UN Number UN1152
Beilstein 1732340
Formula Weight 141.04
Percent Purity 98%
Chemical Name or Material 1,5-Dichloropentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.