missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diiodopentane, 97%, stab. with copper

Product Code. 11463537
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463537 100 g 100g
11453537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463537 Supplier Thermo Scientific Alfa Aesar Supplier No. A17298.22
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 628-77-3 | C5H10I2 | 323.944 g/mol

1,5-Diiodopentane is used as a crosslinking reagent. It is used in the preparation of (iodoalkylaminocarbene)tungsten complexes. It is also used as alkylation to form unsubstituted cyclohexane. It is involved in the preparation of poly(1-vinyl imidazole-co-elthylene glycol methyl ether acrylate electrolytes and used for quasi-solid state dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diiodopentane is used as a crosslinking reagent. It is used in the preparation of (iodoalkylaminocarbene)tungsten complexes. It is also used as alkylation to form unsubstituted cyclohexane. It is involved in the preparation of poly(1-vinyl imidazole-co-elthylene glycol methyl ether acrylate electrolytes and used for quasi-solid state dye-sensitized solar cells.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-77-3
Molecular Formula C5H10I2
Molecular Weight (g/mol) 323.944
MDL Number MFCD00001101
InChI Key IAEOYUUPFYJXHN-UHFFFAOYSA-N
Synonym pentane, 1,5-diiodo, pentamethylene diiodide, 1,5-diiodopentan, pentane,5-diiodo, pubchem3853, acmc-20ap1i, 4-01-00-00317 beilstein handbook reference, iaeoyuupfyjxhn-uhfffaoysa, 1,5-diiodopentane, stab. with copper, 1,5-diiodopentane, contains copper as stabilizer
PubChem CID 12354
IUPAC Name 1,5-diiodopentane
SMILES C(CCI)CCI

Specifications

Density 2.176
Boiling Point 128°C to 130°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6012
Quantity 100 g
Beilstein 1697281
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 323.94
Percent Purity 97%
Chemical Name or Material 1,5-Diiodopentane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.