missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethyl-1H-pyrazole-3-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10405502
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10405502 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10405502 Supplier Thermo Scientific Maybridge Supplier No. CC12602DA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 49783-84-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H7ClN2O
MDL Number MFCD03407950
Quantity 1 g
Synonym 1,5-dimethyl-1h-pyrazole-3-carbonyl chloride, 1h-pyrazole-3-carbonylchloride, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carbonylchloride, 1h-pyrazole-3-carbonyl chloride, 1,5-dimethyl-9ci
InChI Key BADBQYAILRGCBB-UHFFFAOYSA-N
SMILES CN1N=C(C=C1C)C(Cl)=O
IUPAC Name 1,5-dimethylpyrazole-3-carbonyl chloride
Molecular Weight (g/mol) 158.59
PubChem CID 2776364
Formula Weight 158.59
Percent Purity 97%
Chemical Name or Material 1, 5-Dimethyl-1H-pyrazole-3-carbonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.