missing translation for 'onlineSavingsMsg'
Learn More

(1,5-Dimethyl-1H-pyrrol-2-yl)methylamine, 95+%, Thermo Scientific™

Product Code. 10691883
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10691883 1 g 1g
10099561 250 mg 250mg
10357494 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10691883 Supplier Thermo Scientific Maybridge Supplier No. SP01447DA
 Restricted Item
Estimated Shipment: 05-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 118799-24-9
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.187
InChI Key KMUHAJOXPNPERW-UHFFFAOYSA-N
Synonym 1,5-dimethyl-1h-pyrrol-2-yl methylamine, 1,5-dimethyl-1h-pyrrol-2-yl methanamine, 1,5-dimethylpyrrol-2-yl methanamine, 1,5-dimethyl-1h-pyrrol-2-yl methyl amine, 1-1,5-dimethyl-1h-pyrrol-2-yl methanamine, 1,5-dimethylpyrrol-2-yl methylamine, 1-1,5-dimethylpyrrol-2-yl methanamine, 1h-pyrrole-2-methanamine, 1,5-dimethyl, 2-aminomethyl-1,5-dimethyl-1h-pyrrole, 1h-pyrrole-2-methanamine, 1,5-dimethyl-9ci
PubChem CID 737232
IUPAC Name (1,5-dimethylpyrrol-2-yl)methanamine
SMILES CC1=CC=C(N1C)CN

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 124.19
Percent Purity 90%
Chemical Name or Material (1, 5-Dimethyl-1H-pyrrol-2-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.