missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethylhexylamine, TRC

Product Code. 30137890
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137890 10 g 10g
17921295 1 g 1g
17924535 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137890 Supplier Toronto Research Chemicals Supplier No. TRCD47319510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-82-8
Molecular Formula C8 H19 N
Molecular Weight (g/mol) 129.24
Synonym 2-Heptanamine, 6-methyl-, Hexylamine, 1,5-dimethyl- (6CI,7CI,8CI), 6-Methyl-2-heptanamine, (±)-1,5-Dimethylhexylamine, (±)-2-Amino-6-methylheptane, (±)-6-Methyl-2-heptanamine, 1,5-Dimethylhexanamine, 1,5-Dimethylhexylamine, 2-Amino-6-methylheptane, 2-Isooctylamine, 2-Methyl-6-aminoheptane, 6-Amino-2-methylheptane, 6-Methyl-2-heptylamine, Amidrine, Isoctaminum, Octodrin, Octodrine, SKF 51, Vaporpac, dl-2-Amino-6-methylheptane, dl-2-Isooctylamine, α,ε-Dimethylhexylamine
IUPAC Name 6-methylheptan-2-amine
SMILES CC(C)CCCC(C)N

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H19N/c1-7(2)5-4-6-8(3)9/h7-8H,4-6,9H2,1-3H3
Formula Weight 129.1517
Percent Purity >95%
Chemical Name or Material 1,5-Dimethylhexylamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.