missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethylnaphthalene, 99%

Product Code. 11484210
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484210 100 mg 100mg
11494210 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484210 Supplier Thermo Scientific Alfa Aesar Supplier No. A16819.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 571-61-9 | C12H12 | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 571-61-9
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004038
InChI Key SDDBCEWUYXVGCQ-UHFFFAOYSA-N
Synonym naphthalene, 1,5-dimethyl, 1,5-dmn, 1,5-dimethyl-naphthalene, acmc-209lwa, naphthalene,5-dimethyl, 1,5-dimethyl naphthalene, 1,5-dimethylnaphthalene, naphthalene, 1,5-dimethyl-8ci 9ci
PubChem CID 11306
ChEBI CHEBI:48608
IUPAC Name 1,5-dimethylnaphthalene
SMILES CC1=CC=CC2=C(C)C=CC=C12

Specifications

Melting Point 78°C to 82°C
Boiling Point 265°C to 266°C
Odor Odorless
Quantity 100 mg
Beilstein 2039843
Formula Weight 156.23
Percent Purity 99%
Chemical Name or Material 1,5-Dimethylnaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.