missing translation for 'onlineSavingsMsg'
Learn More

Labcon 1.5 mL ViewPoint™ Microcentrifuge Thermochromic Tubes
GREENER_CHOICE

Product Code. 16649832
Click to view available options
Capacity (Metric):
1.5 mL
Unit Size:
Pack of 500
This item is not returnable. View return policy

Product Code. 16649832

Brand: Labcon 30058400009

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C

  • ViewPoint™ Thermochromic
  • Real-time temperature signaling
  • Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C
  • 1.5 mL
  • With flat easy open attached snap caps; side graduations, and frosted writing surface 
  • Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Sperm Motility Assay (SMA) Mouse Embryo Assay (MEA) validated safe for cell culture
  • Polypropylene USP Class VI resin
  • Ultra high centrifuging to 40,000 x G
  • Freezable to -80°C
  • Made in USA

Specifications

Cap Style Snap Cap
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Clear to Blue (Color change at 0°C to 4°C)
Material Polypropylene Resin
For Use With (Application) Centrifugation
Packaging 100 / resealable bag/5 bags / Pk./2 Pk. / case
Temperature Range (Metric) Freezable to -80°C
Clean Claims Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic), Sperm Motility Assay (SMA) Mouse Embryo Assay (MEA) validated safe for cell culture
Sterility Non-sterile
Shipping Condition RT
Product Line ViewPoint
Graduated Yes
Max. RCF 40,000 RCF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.