missing translation for 'onlineSavingsMsg'
Learn More

1,5-Pentanediol, 98%

Product Code. 10071320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Metal drum
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10071320 500 g Glass bottle 500g
10656522 100 g Glass bottle 100g
10521571 1 kg Glass bottle 1kg
11305932 5 kg Metal drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10071320 Supplier Thermo Scientific Acros Supplier No. 129955000
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-29-5 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-29-5
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002978
InChI Key ALQSHHUCVQOPAS-UHFFFAOYSA-N
Synonym 1,5-pentanediol, 1,5-dihydroxypentane, pentylene glycol, pentamethylene glycol, 1,5-pentylene glycol, 1,5 pentanediol, 1,5-pentamethylene glycol, alpha,omega-pentanediol, unii-07uxz0scst, 1,5-pentandiol
PubChem CID 8105
IUPAC Name pentane-1,5-diol
SMILES OCCCCCO

Specifications

Melting Point -18.0°C
Color Colorless
Density 0.9940g/mL
Boiling Point 242.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO(CH2)5OH
Refractive Index 1.4490 to 1.4510
Quantity 500 g
Beilstein 01,481
Merck Index 15,7229
Specific Gravity 0.994
Solubility Information Solubility in water: miscible. Other solubilities: limited solubility in benzene, trichloroethylene,, petroleum ether, heptane, miscible with alcohol, acetone and ethyl acetate, 11 g/110g ether (25°C)
Viscosity 77 mPa.s (30°C)
Formula Weight 104.15
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material 1, 5-Pentanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.