missing translation for 'onlineSavingsMsg'
Learn More

1,5-Pentanedithiol, TRC

Product Code. 30420678
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420678 1 g
30413477 500 mg
30409175 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420678 Supplier Toronto Research Chemicals Supplier No. TRCP2274801G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-98-3
Molecular Formula C5 H12 S2
Molecular Weight (g/mol) 136.28
Synonym 1,5-Dimercaptopentane, 1,5-Dithiolpentane, NSC 51699
IUPAC Name pentane-1,5-dithiol
SMILES SCCCCCS

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H12S2/c6-4-2-1-3-5-7/h6-7H,1-5H2
Formula Weight 136.038
Chemical Name or Material 1,5-Pentanedithiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.