missing translation for 'onlineSavingsMsg'
Learn More

1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid, Thermo Scientific™

Product Code. 10883951
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10883951 250 mg Amber Glass Bottle 250mg
10646430 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10883951 Supplier Thermo Scientific Maybridge Supplier No. CC73101CB
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 950603-35-7
Molecular Formula C9H10F3N3O2S
Molecular Weight (g/mol) 281.25
MDL Number MFCD11101440
InChI Key PWQMNLMMMCREOV-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl-1,3,4-thiadiazol-2-yl piperidine-4-carboxylic acid
PubChem CID 33589561
IUPAC Name 1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid
SMILES OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 281.25
Percent Purity 97%
Chemical Name or Material 1-[5-(trifluoromethyl)-1, 3, 4-thiadiazol-2-yl]piperidine-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.