missing translation for 'onlineSavingsMsg'
Learn More

1,6-Anhydro-beta-D-galactose, TRC

Product Code. 17975005
Change view
Click to view available options
Quantity:
25 mg
500 mg
1 g
Unit Size:
1g
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17975005 500 mg 500mg
17921605 25 mg 25mg
17972145 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17975005 Supplier Toronto Research Chemicals Supplier No. TRCA641000500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 644-76-8
Molecular Formula C6 H10 O5
Molecular Weight (g/mol) 162.14
Synonym Galactopyranose, 1,6-anhydro-, β-D- (6CI,8CI), 1,6-Anhydro-β-D-galactopyranose, 6,8-Dioxabicyclo[3.2.1]octane, β-D-galactopyranose deriv., D-Galactosan, Galactosan, Levogalactosan, NSC 1376, Galactopyranose, 1,6-anhydro-, β-D- (6CI,8CI), 1,6-Anhydro-β-D-galactopyranose, 6,8-Dioxabicyclo[3.2.1]octane, β-D-galactopyranose deriv., D-Galactosan, Galactosan, Levogalactosan, NSC 1376
IUPAC Name (1R,2R,3S,4R,5R)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
SMILES O[C@H]1[C@H]2CO[C@H](O2)[C@H](O)[C@H]1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H10O5/c7-3-2-1-10-6(11-2)5(9)4(3)8/h2-9H,1H2/t2-,3+,4+,5-,6-/m1/s1
Formula Weight 162.0528
Percent Purity >95%
Chemical Name or Material 1,6-Anhydro-D-galactose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.