missing translation for 'onlineSavingsMsg'
Learn More

1,6-Anhydro-beta-D-mannopyranose, TRC

Product Code. 30136303
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136303 10 g 10g
17917604 500 mg 500mg
17919764 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136303 Supplier Toronto Research Chemicals Supplier No. TRCA65250010G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14168-65-1
Molecular Formula C6 H10 O5
Molecular Weight (g/mol) 162.14
Synonym Mannopyranose, 1,6-anhydro-, β-D- (6CI,8CI), 1,6-Anhydro-β-D-mannopyranose, 6,8-Dioxabicyclo[3.2.1]octane, β-D-mannopyranose deriv., 1,6-Anhydro-D-mannose, 1,6-Anhydromannose, Mannosan, NSC 226600
IUPAC Name (1R,2S,3S,4S,5R)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
SMILES O[C@@H]1[C@H]2CO[C@H](O2)[C@@H](O)[C@H]1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H10O5/c7-3-2-1-10-6(11-2)5(9)4(3)8/h2-9H,1H2/t2-,3-,4+,5+,6-/m1/s1
Formula Weight 162.0528
Percent Purity >95%
Chemical Name or Material 1,6-Anhydro-beta-D-mannopyranose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.