missing translation for 'onlineSavingsMsg'
Learn More

1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate, 98+%, Thermo Scientific™

Product Code. 10272562 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
Product Code. Quantity Packaging unitSize
10272562 500 mg Glass bottle 500mg
10120132 100 mg Glass bottle 100mg
2 options
This item is not returnable. View return policy

Product Code. 10272562

Brand: Thermo Scientific Acros 353685000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13242-55-2
Molecular Formula C12H16O8
Molecular Weight (g/mol) 288.25
MDL Number MFCD00063247
InChI Key BAKQMOSGYGQJOJ-UHFFFAOYNA-N
Synonym triacetyllevoglucosan, 1,6-anhydro-, a-d-glucopyranose 2,3,4-triacetate, 2,3,4-tri-o-acetyl-1,6-anhydro-, a-d-glucopyranose, 2r,3s,4r,5r-2,3-bis acetyloxy-6,8-dioxabicyclo 3.2.1 octan-4-yl acetate
PubChem CID 134159065
IUPAC Name [(1S,2S,3S,4S,5R)-3,4-diacetyloxy-6,8-dioxabicyclo[3.2.1]octan-2-yl] acetate
SMILES CC(=O)OC1C2COC(O2)C(OC(C)=O)C1OC(C)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 111.0°C to 112.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 500 mg
Specific Rotation − 66.00
Solubility Information Solubility in water: soluble.
Formula Weight 288.26
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.