missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,6-Bis(mesyloxy)hexane, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 1,6-Bis(mesyloxy)hexane |
| CAS | 4239-24-1 |
| Molecular Formula | C8H18O6S2 |
| Quantity | 1 g |
| Synonym | Methanesulfonic Acid Hexamethylene Ester, 1,6-Hexamethylene Dimesylate, 6-Methylsulfonyloxyhexyl Methylsulfonate, Hexasulfan, Hexasulphan, 1,6-Hexanediol Dimethanesulfonate |
| InChI Formula | InChI=1S/C8H18O6S2/c1-15(9,10)13-7-5-3-4-6-8-14-16(2,11)12/h3-8H2,1-2H3 |
| SMILES | CS(=O)(=O)OCCCCCCOS(=O)(=O)C |
| IUPAC Name | 6-methylsulfonyloxyhexyl methanesulfonate |
| Molecular Weight (g/mol) | 274.35 |
| Formula Weight | 274.0545 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction