missing translation for 'onlineSavingsMsg'
Learn More

1-(6-Chloro-3-pyridinyl)-1-ethanone, 90%, Thermo Scientific™

Product Code. 10069341
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10069341 10 g Amber Glass Bottle 10g
10662063 1 g Amber Glass Bottle 1g
10174244 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10069341 Supplier Thermo Scientific Maybridge Supplier No. MO00162EA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 55676-22-7
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD03407343
InChI Key UXSNZYGTQTXRAD-UHFFFAOYSA-N
Synonym 1-6-chloropyridin-3-yl ethanone, 2-chloro-5-acetylpyridine, 3-acetyl-6-chloropyridine, 5-acetyl-2-chloropyridine, 1-6-chloropyridin-3-yl ethan-1-one, 1-6-chloro-3-pyridinyl-1-ethanone, 6-chloro-3-acetylpyridine, 2-chloro-5-acetyl-pyridine, 1-6-chloro-3-pyridinyl ethanone, 1-6-chloro-pyridin-3-yl-ethanone
PubChem CID 2779698
IUPAC Name 1-(6-chloropyridin-3-yl)ethanone
SMILES CC(=O)C1=CN=C(Cl)C=C1

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 155.58
Percent Purity 90%
Chemical Name or Material 1-(6-Chloro-3-pyridinyl)-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.